Learn More
Leinco Technologies Inc Anti-Mouse CD172a (SIRPα, SHPS-1, BIT, P84, PTPNS1) In Vivo GOLD; Clone P84; Isotype: Rat IgG1 κ
SDP
Supplier:  Leinco Technologies Inc P380100 MG
Encompass_Preferred
CD172a and its ligand CD47 are thought to modify neural synaptic activity and regulate the phagocytic activities of macrophages via the mediation of bi-directional signaling.
Catalog No. 50-190-6604