Learn More
Leinco Technologies Inc Anti-Mouse CD172a (SIRPα, SHPS-1, BIT, P84, PTPNS1) In Vivo PLATINUM; Clone P84; Isotype: Rat IgG1 κ
SDP
Supplier:  Leinco Technologies Inc P6805.0 MG
Encompass_Preferred
Specificity: Clone P84 recognizes an epitope on mouse CD172a. Antigen Distribution: CD172a is expressed on monocytes, macrophages, myeloid cells, and neuronal tissue. Background: CD172a is a regulatory membrane type I glycoprotein from the SIRP family. CD172a and its ligand CD47 are thought to modify neural synaptic activity and regulate the phagocytic activities of macrophages via the mediation of bi-directional signaling. Applications: (WB, B) Reported applications include blocking the interaction of CD172a with CD47, and in vivo blocking of dendritic cell migration. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6621