Learn More
Leinco Technologies Inc Anti-Mouse CD223 (LAG-3) In Vivo GOLD; Clone C9B7W; Isotype: Rat IgG1
SDP
Supplier:  Leinco Technologies Inc L3065.0 MG
Encompass_Preferred
Specificity: Clone C9B7W recognizes & specifically binds to an epitope in the D2 domain of CD223. Antigen Distribution: CD223 is expressed on T regulatory cells, activated T cells & NK cells. Background: CD223 is a 70-kD, type-I transmembrane glycoprotein within the Ig superfamily & is structurally homologous to CD4. CD223 binds MHC class II molecules. Applications: (IP, WB, FA) C9B7W has been shown to inhibit the suppressor function of T regulatory cells in vitro & in vivo. Clone C9B7W blocks the CD223-mediated inhibition of IL-2 production by a T-cell hybridoma responding to antigen. C9B7W is unable to block the binding of MHC class II-IgG2a fusion protein to CD223. Leinco’s In vivo GOLDTM & In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques & are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6521