Learn More
Leinco Technologies Inc Anti-Mouse CD278 (ICOS, H4, ALIM) In Vivo PLATINUM; Clone 7E.17G9; Isotype: Rat IgG2b
SDP
Supplier:  Leinco Technologies Inc C285125 MG
Encompass_Preferred
Specificity: This antibody recognizes an epitope on mouse CD278, a 47-57 kD protein. Antigen Distribution: ICOS is expressed on activated T cells and APCs including splenic B cells, dendritic cells, and macrophages. Background: It enhances all basic T cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B cells. ICOS signaling is an important regulator for T cell homeostasis. Applications: (WB, B, FA) This clone has been shown to block binding of ICOSL to ICOS. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6002