Learn More
Leinco Technologies Inc Anti-Mouse PD-1 (Programmed Death-1, CD279, PD 1) In Vivo GOLD; Clone 29F.1A12; Isotype: Rat IgG2a κ
SDP
Supplier:  Leinco Technologies Inc P3775.0 MG
Encompass_Preferred
Specificity: Clone 29F.1A12 recognizes an epitope on mouse PD-1. Antigen distribution: PD-1 is expressed on a subset of CD4-CD8- thymocytes, and on activated T and B cells. Background: PD-1 is a 50-55 kD member of the Ig superfamily and is suspected to play a role in lymphocyte clonal selection and peripheral tolerance. The ligands of PD-1 are PD-L1 (B7-H1) and PD-L2 (B7-DC). The ligands are members of the B7 Ig superfamily. Applications: (WB, B) Similar to clones RMP1-14 and J43, clone 29F.1A12 has been shown to block the binding of PD-1 to its ligands In vivo. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6586