Learn More
Leinco Technologies Inc Anti-Mouse CD3e (T3, CD3, CD3e) In Vivo GOLD; Clone 145-2C11; Isotype: Armenian Hamster IgG
SDP
Supplier:  Leinco Technologies Inc C17585.0 MG
Encompass_Preferred
Specificity: Clone 145-2C11 reacts with mouse CD3ε. Antigen Distribution: CD3ε is primarily expressed on T cells, NK-T cells, and at different levels on thymocytes during T cell differentiation. Background: CD3ε is a 20kDa transmembrane cell-surface protein that belongs to the Ig superfamily. Applications: (IP, B, FC, WB) Some reported applications for the 145-2C11 antibody are In vitro blocking and activation assays, apoptosis induction, In vivo T cell depletion, and immunohistochemical staining. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5849