Learn More
Leinco Technologies Inc Anti-Mouse CD3ε (CD3, T3) In Vivo PLATINUM; Clone 500 A2; Isotype: Syrian Hamster IgG2a κ
SDP
Supplier:  Leinco Technologies Inc C28341.0 MG
Encompass_Preferred
Specificity: Clone 500 A2 recognizes an epitope on mouse CD3ε. Antigen Distribution: CD3ε is expressed on differentiation-dependent thymocytes, mature T cells and NK-T cells. Background: CD3ε is a 20 kDa transmembrane protein and is a member of the Ig superfamily. It is one of five polypeptide chains that form the TCR complex by associating with the CD3δ, γ and ζ chains in addition to the TCR α/β or γ/δ chains. CD3 is involved in TCR signaling, T lymphocyte activation, and antigen recognition. Applications: (FA) The binding of Hamster Anti-Mouse CD3ε (Clone 500 A2) to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5945