Learn More
Leinco Technologies Inc Anti-Mouse L-Selectin (CD62L) In vivo PLATINUM; Clone MEL-14; Isotype: Rat IgG2a κ
SDP
Supplier:  Leinco Technologies Inc C61185.0 MG
Encompass_Preferred
Specificity: Clone MEL-14 recognizes an epitope on mouse CD62L. Antigen Distribution: L-selectin is expressed on the majority of B and naïve T cells, a subset of memory T cells, monocytes, granulocytes, most thymocytes, and a subset of NK cells. Background: CD62L is a 74-95 kD glycoprotein and is a member of the selectin family. Applications: (IP, WB, B, FC) MEL-14 is reported to block adhesion In vivo and In vitro and is used in immunohistochemical staining. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6169