Learn More
Leinco Technologies Inc Anti-Mouse CD8a (Ly 2.2, Ly-2, Ly-35, Ly-B, Lyt-2, Lyt2) In Vivo GOLD; Clone 2.43; Isotype: Rat IgG2b
SDP
Supplier:  Leinco Technologies Inc C38025 MG
Encompass_Preferred
Specificity: Clone 2.43 recognizes an epitope on mouse CD8a. Antigen Distribution: CD8a is present on the surface of most thymocytes and a subpopulation of mature T-lymphocytes which include most T suppressor/cytotoxic-cells. Background: CD8 participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Applications: (WB, FA) Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6120