Learn More
Leinco Technologies Inc Anti-Mouse CD8b.2 (Lyt-3.2, Ly-3.2) In Vivo GOLD; Clone 53-5.8; Isotype: Rat IgG1 κ
SDP
Supplier:  Leinco Technologies Inc C283225 MG
Encompass_Preferred
Specificity: Clone 53-5.8 recognizes an epitope on mouse CD8b.2. Antigen Distribution: CD8b.2 is expressed on the majority of thymocytes and cytotoxic T cell subsets. Background: It is a member of the Ig superfamily and is expressed as a heterodimer with the CD8α chain on a subset of MHC class I-restricted T cells and most thymocytes. Applications: (WB, FA) It is specific for Ly-3.2 and has low reactivity with Ly-3.1. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5937