Learn More
Seracare Life Sciences Inc ANTI-MS G+M (H+L) AB BIOTIN
Catalog No. 5067278
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG and IgM, heavy and light chains (whole IgA, IgG, IgM) made in Goat and biotinylated. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-672-78 Supplier Seracare Life Sciences Inc Supplier No. 52600050
Affinity purified polyclonal antibody to Mouse IgG and IgM, heavy and light chains (whole IgA, IgG, IgM) made in Goat and biotinylated. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.