Learn More
Seracare Life Sciences Inc ANTI-MS G+M (H+L) AB HSA FITC
Supplier:  Seracare Life Sciences Inc 52300309
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Mouse IgG and IgM, both heavy and light chains (whole IgG and IgM) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross-adsorbed with human serum.
Catalog No. 50-668-95