Learn More
Leinco Technologies Inc Anti-Mouse GM-CSF (CSF-2, MGI-1GM, Pluripoietin-Alpha) In Vivo PLATINUM; Clone MP1-22E9; Isotype: Rat IgG2a
SDP
Supplier:  Leinco Technologies Inc G7415.0 MG
Encompass_Preferred
Specificity: Clone MP122E9 recognizes an epitope on mouse GM-CSF. Antigen distribution: GM-CSF is produced by T cells, macrophages, fibroblasts and endothelial cells. Background: Granulocyte-Macrophage Colony Stimulating Factor is a 22 kD, pleiotropic cytokine that controls the production and function of blood cells. GM-CSF stimulates stem cells to produce granulocytes and monocytes. GM-CSF has a critical role in regulation of surfactant homeostasis and alveolar macrophage innate immune functions in the lung. Applications: (WB) This antibody shows no cross-reactivity with rhGM-CSF. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6315