Learn More
Leinco Technologies Inc Anti-Mouse IFNß (HDb-4A7) In Vivo GOLD; Clone HDß-4A7; Isotype: Mouse IgG2a
SDP
Supplier:  Leinco Technologies Inc I11821.0 MG
Encompass_Preferred
Specificity: Clone HDß-4A7 binds specifically to murine IFNß. Antigen distribution: IFNß is expressed on fibroblasts and epithelial cells. Background: IFN-β is a 26-35 kD glycoprotein type I IFN and is involved in the innate immune system, being released non-specifically during the early stage of infection in response to stimuli such as viruses or bacterial components. IFN-β is the therapeutic target for treating multiple sclerosis. Applictions: (WB, B, FA) HDb-4A7 binds both recombinant and natural forms of IFNb. Clone HDß-4A7blocks In vivo IFNb leading to increased the lethality of mice infected with West Nile Virus, like susceptibility of IFNb-/- mice. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6408