Learn More
Leinco Technologies Inc Anti-Mouse IFNγ (Interferon Gamma, Immune Interferon, Type II Interferon, T Cell Interferon, MAF, IFNG, IFG, IFN-y) In Vivo GOLD; Clone H22; Isotype: Armenian Hamster IgG
SDP
Supplier:  Leinco Technologies Inc I43825 MG
Encompass_Preferred
Specificity: Clone H22 recognizes mouse IFNγ. Cell sources: IFNγ is produced predominantly by NK and NKT cells as part of the innate immune response, in addition to CD4 and CD8 cytotoxic T lymphocyte (CTL) effector T cells once antigen-specific immunity develops. Background: IFNγ is a pleiotropic cytokine of 15-17 kD that is the only member of the type II class of interferons. This biologically active dimer is vital for innate and adaptive immunity against viral and intracellular bacterial infections, and for tumor control. Applications: (IP, WB) H22 may inhibit the In vitro and In vivo activities of mouse IFNγ. H22 does not react with mouse IFN-α, IFN-β, IL-6, or human IFNγ. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6482