Learn More
Seracare Life Sciences Inc ANTI-MS IGA+IGG+IGM(H+L)AB HSA
Supplier:  Seracare Life Sciences Inc 52100187
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgA, IgG, and IgM, both heavy and light chains (whole IgA, IgG, and IgM) made in Goat. Product has been cross-adsorbed with human serum to reduce cross-reactivity to human immunoglobulin.
Catalog No. 50-668-27