Learn More
Seracare Life Sciences Inc ANTI-MS IGG (G) AB HSA DL488
Catalog No. 5067083
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG heavy chains (gamma) made in Goat and labeled with DyLight 488. Cross-reactivity with human serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-670-83 Supplier Seracare Life Sciences Inc Supplier No. 52300389
Affinity purified polyclonal antibody to Mouse IgG heavy chains (gamma) made in Goat and labeled with DyLight 488. Cross-reactivity with human serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).