Learn More
Seracare Life Sciences Inc ANTI-MS IGG (G) AB HSA FITC
Supplier:  Seracare Life Sciences Inc 52300305
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG heavy chains (gamma) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin.
Catalog No. 50-668-91