Learn More
Seracare Life Sciences Inc ANTI-MS IGG(G) F(AB )2 FRAGHSA
Catalog No. 5067308
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Mouse IgG heavy chains (gamma). Made in Goat. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-673-08 Supplier Seracare Life Sciences Inc Supplier No. 52100302
Affinity purified polyclonal antibody, F(ab)2 fragment, to Mouse IgG heavy chains (gamma). Made in Goat. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.