Learn More
Seracare Life Sciences Inc ANTI-MS IGG (G) F(AB )2 HRP
Supplier:  Seracare Life Sciences Inc 52200394
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Mouse IgG heavy chains (gamma). Made in Goat and labeled with horseradish peroxidase. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-65
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'