Learn More
Seracare Life Sciences Inc ANTI-MS IGG(H+L) AB HSA BIOTIN
Catalog No. 5067303
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-673-03 Supplier Seracare Life Sciences Inc Supplier No. 52600061
Affinity purified polyclonal antibody to Mouse IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.