Learn More
Seracare Life Sciences Inc ANTI-MS IGG (H+L) AB RB+HSA AP
Supplier:  Seracare Life Sciences Inc 52200312
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. Product has been cross-adsorbed to rabbit immunoglobulin and human serum to minimize cross-reactivity. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-13