Learn More
Seracare Life Sciences Inc ANTI-MS IGG(H+L)AB RB+HSAFITC
Supplier:  Seracare Life Sciences Inc 52300311
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross-adsorbed with human serum.
Catalog No. 50-668-97