Learn More
Seracare Life Sciences Inc ANTI-MS IGG (H+L) AB RESERVEAP
Supplier:  Seracare Life Sciences Inc 52200357
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. ReserveAP conjugates represent the latest advances in KPL's conjugation chemistry and are more sensitive than KPL's standard AP conjugates. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-19