Learn More
Seracare Life Sciences Inc ANTI-MS IGG(H+L) AB RT+HSA HRP
Supplier:  Seracare Life Sciences Inc 52200287
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to rat immunoglobulin and human serum to minimize cross-reactivity.
Catalog No. 50-669-42