Learn More
Seracare Life Sciences Inc ANTI-MS IGG (H+L) ABRB+HSA HRP
Supplier:  Seracare Life Sciences Inc 52200288
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to rabbit immunoglobulin and human serum to minimize cross-reactivity.
Catalog No. 50-669-43