Learn More
Seracare Life Sciences Inc ANTI-MS IGG(H+L) F(AB )2 FITC
Catalog No. 5067360
Encompass_Preferred
Affinity purified polyclonal antibody, F(abAE)2 fragment, to Mouse IgG, heavy and light chains (whole IgG). Made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-673-60 Supplier Seracare Life Sciences Inc Supplier No. 52300446
Affinity purified polyclonal antibody, F(abAE)2 fragment, to Mouse IgG, heavy and light chains (whole IgG). Made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.