Learn More
Seracare Life Sciences Inc ANTI-MS IGG (H+L) HSA AB DL488
Catalog No. 5066951
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Cross-reactivity with human serum has been minimized with affinity procedures.
This item is not returnable. View return policy
Catalog No. 50-669-51 Supplier Seracare Life Sciences Inc Supplier No. 52300341
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Cross-reactivity with human serum has been minimized with affinity procedures.