Learn More
Seracare Life Sciences Inc ANTI-MS IGG(H+L) HSA AB DL680
Supplier:  Seracare Life Sciences Inc 52300344
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 680. Cross-reactivity with human serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-669-63