Learn More
Seracare Life Sciences Inc ANTI-MS IGG+IGM(H+L)AB HSA HRP
Supplier:  Seracare Life Sciences Inc 52200343
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG and IgM, heavy and light chains (whole IgG and IgM) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-08