Learn More
Seracare Life Sciences Inc ANTI-MS IGG+IGM (H+L) AB HSA
Supplier:  Seracare Life Sciences Inc 52100188
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG and IgM, both heavy and light chains (whole IgG and IgM) made in Goat. Product has been cross-adsorbed with human serum to reduce cross-reactivity to human immunoglobulin.
Catalog No. 50-668-28