Learn More
Leinco Technologies Inc Anti-Mouse Ly-6G (Lymphocyte antigen 6 complex, locus G) In Vivo PLATINUM; Clone 1A8; Isotype: Rat IgG2a
SDP
Supplier:  Leinco Technologies Inc L3055.0 MG
Encompass_Preferred
Specificity: Clone 1A8 recognizes an epitope on mouse Ly-6G. Antigen Distribution: Ly-6G is expressed on the majority of myeloid cells in bone marrow and peripheral granulocytes. Background: Ly-6G is a 21-25 kD GPI-anchored protein. Applications: (IHC (NBF/Par.), IHC (Frozen), FC, WB, In vivo Depletion) Clone RB6-8C5 recognizes both Ly-6G and Ly-6C, while clone 1A8 recognizes Ly-6G alone with no reported cross-reactivity with Ly6C. RB6-8C5 binds with high affinity to mouse Ly-6G and to a lesser extent, Ly-6C. RB6-8C5 inhibits the binding of 1A8, and RB6-8C5 is able to stain in the presence of clone HK1.4. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6516