Learn More
Leinco Technologies Inc Anti-Mouse MHC Class I (Class I H-2Kd) In Vivo GOLD; Clone A4C8.1-Do9; Isotype: Mouse IgG1 κ
SDP
Supplier:  Leinco Technologies Inc H14625 MG
Encompass_Preferred
Specificity: Clone A4C8.1-Do9 recognizes an epitope on mouse Class I H-2Kd. Antigen distribution: Class I H-2Kd is found on the cells from mice of the H-2Kd haplotype. They may occur on platelets, but not on red blood cells. Background: H-2Kd is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Applications: (WB) Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6321