Learn More
Leinco Technologies Inc Anti-Mouse TNF R2 (Tumor Necrosis Factor Receptor II, TNF RII, p75, CD120b, TBPII, TNF-R75, TNFBR, TNFR2, TNFR80, p75TNFR) In Vivo GOLD; Clone TR75-54.7; Isotype: Armenian Hamster IgG
SDP
Supplier:  Leinco Technologies Inc T9541.0 MG
Encompass_Preferred
Specificity: Clone TR75-54.7 recognizes mouse TNFR2. Antigen distribution: TNFR2 is expressed by a variety of cell types including hematopoietic cells such as macrophages, neutrophils, lymphocytes, thymocytes & mast cells. Background: TNFR2 is a 75 kD type I transmembrane protein that binds both TNF-α & TNF-β (LT-α). Together with TRAF1 & TRAF2, the receptor crosslinking induced by TNF-α or TNF-β trimers is vital for signal transduction that may lead to apoptosis, NF-kB activation, increased expression of proinflammatory genes, tumor necrosis, & cell differentiation. Applications: (IP, WB, B) Clone TR75-54.7 has been reported to block ligand-induced receptor signaling. Leinco’s In vivo GOLDTM & In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques & are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6727