Learn More
Mabtech Inc Anti-mouse IFN-gamma mAb AN18, purified, 1 mg
Catalog No. NC9963961
Encompass
This monoclonal antibody enables sensitive and specific detection of mouse IFN-gamma in immunoassays such as ELISA and ELISpot.
Catalog No. NC9963961 Supplier Mabtech Inc Supplier No. 332131000
This monoclonal antibody enables sensitive and specific detection of mouse IFN-gamma in immunoassays such as ELISA and ELISpot.