Learn More
Seracare Life Sciences Inc ANTI-MSIG(A+G+M)(H+L)ABHSA HRP
Supplier:  Seracare Life Sciences Inc 52200342
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgA, IgG, and IgM, heavy and light chains (whole IgA, IgG, IgM) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-07