Learn More
Seracare Life Sciences Inc ANTI-MSIGG (H+L) AB RT+HSA AP
Catalog No. 5067012
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. Product has been cross-adsorbed to rat immunoglobulin and human serum to minimize cross-reactivity. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-670-12 Supplier Seracare Life Sciences Inc Supplier No. 52200311
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. Product has been cross-adsorbed to rat immunoglobulin and human serum to minimize cross-reactivity. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.