Learn More
Seracare Life Sciences Inc ANTI-MSIGG(H+L)F(AB )2 FRAGHSA
Supplier:  Seracare Life Sciences Inc 52100303
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Mouse IgG, heavy and light chains (whole IgG). Made in Goat. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-09