Learn More
Seracare Life Sciences Inc ANTI-MSIGG(H+L) F(AB )2 HSA AP
Supplier:  Seracare Life Sciences Inc 52200399
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment to Mouse IgG, heavy and light chains (whole IgG).Made in Goat and labeled with alkaline phosphatase. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-72