Learn More
Seracare Life Sciences Inc ANTI-MSIGG(H+L)RBSA AB HSA CY3
Supplier:  Seracare Life Sciences Inc 52300366
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with Cy3. Cross-reactivity with human and rabbit serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-49