Learn More
BioVendor, LLC Anti-Mullerian Hormone Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL012R
Encompass
Anti-M¿llerian Hormone (AMH) is a glycoprotein belonging to the transforming growth factors (TGF-P). In females, AMH is secreted by granulosa cells of small follicles in the ovary. Serum AMH levels strongly correlate with the number of growing follicles. Serum AMH levels are used in individualized follicle-stimulating hormone dosing protocols and may predict the risk of poor response or ovarian hyperstimulation syndrome.3 Serum concentrations of AMH gradually decrease and fall below detectable levels in menopause. AMH is the best current available measure of ovarian reserve for different clinical conditions. One kit (96 wells). Type: Sandwich ELISA. Sample type: serum, plasma. Sample requirement: 30 ¿l/well. Calibration range: 0.094-3ng/ml. Limit of detection: 25pg/ml. Other names: AMH, Muellerian-inhibiting substance.
Catalog No. 50-240-6372