Learn More
Bioworld Anti-Myc Tag (Myc.A7 / M19) Antibody (Mouse Monoclonal IgG), 100 μg
Supplier:  Bioworld 203110061
Encompass_Preferred
Anti-Myc Tag (Myc.A7 / M19) Antibody (Mouse Monoclonal IgG)Cat#20311006DetailsHost:MouseIsotype:IgGClass:MonoclonalImmunogen:Synthetic peptide corresponding toMyc Tag (Myc.A7 / M19)Conjugate:UnconjugatedForm InformationForm:LiquidConcentration:2 mg/mLPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:Shipped at 4°C. Upon delivery aliquot and store at -20°C. Avoid freeze / thaw cycles.Usage RecommendationsWestern Blot (WB):1:1000-1:3000ELISA:1:150-1:400Immunoprecipitation (IP):1:5000 - 1:2000
Catalog No. 50-197-997