Learn More
Immunology Consultants anti-Mycoplasma pneumoniae Antibody (Mouse - Clone #1C1) - Monoclonal
SDP
Catalog No. 5000435289
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of Mycoplasma pneumoniae, a major respiratory pathogen. Clone 1C1 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays, enabling accurate detection in clinical research samples, culture supernatants, and purified antigen preparations. MMPP‑55A‑1C1 is ideal for diagnostic assay development, microbial surveillance, and studies examining M. pneumoniae infection mechanisms. Its reproducible performance ensures dependable antigen recognition across repeated assays.
Catalog No. 50-004-35289 Supplier Immunology Consultants Supplier No. MMPP55A1C1
This monoclonal antibody provides sensitive, specific detection of Mycoplasma pneumoniae, a major respiratory pathogen. Clone 1C1 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays, enabling accurate detection in clinical research samples, culture supernatants, and purified antigen preparations. MMPP‑55A‑1C1 is ideal for diagnostic assay development, microbial surveillance, and studies examining M. pneumoniae infection mechanisms. Its reproducible performance ensures dependable antigen recognition across repeated assays.