Learn More
Immunology Consultants anti-Mycoplasma pneumoniae Antibody (Mouse - Clone #5E12) - Monoclonal
SDP
Catalog No. 5000435543
Encompass_Preferred
Clone 5E12 targets a distinct M. pneumoniae epitope, providing complementary detection to Clone 1C1. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MMPP‑55A‑5E12 is well suited for diagnostic development, pathogen monitoring, and research on respiratory infection dynamics. Its consistent performance ensures reliable detection across diverse sample types.
Catalog No. 50-004-35543 Supplier Immunology Consultants Supplier No. MMPP55A5E12
Clone 5E12 targets a distinct M. pneumoniae epitope, providing complementary detection to Clone 1C1. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MMPP‑55A‑5E12 is well suited for diagnostic development, pathogen monitoring, and research on respiratory infection dynamics. Its consistent performance ensures reliable detection across diverse sample types.