Learn More
Immunology Consultants anti-Mycoplasma pneumoniae Antibody (Mouse - Clone #7A12) - Monoclonal
SDP
Supplier:  Immunology Consultants MMPP55A7A12
Encompass_Preferred
This monoclonal antibody recognizes a unique antigenic determinant on M. pneumoniae, enabling precise detection in immunoassays. Clone 7A12 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture formats. MMPP‑55A‑7A12 is ideal for diagnostic assay development, microbial surveillance, and studies examining host–pathogen interactions. Its reproducible performance ensures dependable antigen detection across repeated assays and complex sample matrices.
Catalog No. 50-004-35511