Learn More
Biolegend Anti-Myosin Phosphatase
Supplier:  Biolegend 925101
Encompass_Preferred
Anti-Myosin Phosphatase [Poly19251]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, Dog, Mouse, Rat, Rodent; Apps: WB; Size: 100 uL
Catalog No. 50-207-5263