Learn More
Chondrex Inc ANTI-NC1 COL4 IGG2A, 0.1 MG
Catalog No. 501526893
Encompass_Preferred
Rat monoclonal IgG2a antibody, clone H11. Immunogen: Human NC1 domain of alpha 1 chain of type IV collagen (KKPTPSTL). Form: 100 µL of 1 mg/ml solution in PBS. Reactivity: Human, canine, mouse, and rat NC1 domain of the alpha 1 chain of type IV collagen. Application: IHC, Immunoblotting.
Catalog No. 50-152-6893 Supplier Chondrex Inc Supplier No. 7070
Add to Cart
Add to Cart
Rat monoclonal IgG2a antibody, clone H11. Immunogen: Human NC1 domain of alpha 1 chain of type IV collagen (KKPTPSTL). Form: 100 µL of 1 mg/ml solution in PBS. Reactivity: Human, canine, mouse, and rat NC1 domain of the alpha 1 chain of type IV collagen. Application: IHC, Immunoblotting.