Learn More
Sino Biological Anti-NCAM-1/CD56 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750500
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NCAM-1/CD56 (rh NCAM-1/CD56; Catalog#10673-H08H; NP_001070150.1; Met1-Pro603). NCAM-1/CD56 specific IgG was purified by Human NCAM-1/CD56 affinity chromatography.
Catalog No. 50-007-50500 Supplier Sino Biological Supplier No. 10673T24100
Produced in rabbits immunized with purified, recombinant Human NCAM-1/CD56 (rh NCAM-1/CD56; Catalog#10673-H08H; NP_001070150.1; Met1-Pro603). NCAM-1/CD56 specific IgG was purified by Human NCAM-1/CD56 affinity chromatography.