Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-NECTIN4 antibody produced in rabbit, 100 uL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HPA010775100UL
Encompass
Anti-PVRL4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project. Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues.
Catalog No. NC1925029