Learn More
Biolegend Anti-Neurofilament M (NF-M)
Supplier:  Biolegend 841001
Encompass_Preferred
Anti-Neurofilament M (NF-M) [Poly28410]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, Rat, Mouse,; Apps: IHC-P, WB; Size: 100 uL
Catalog No. 50-103-0056